Assay Details
Gene Symbol: CUNH9orf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013072658.2 | XP_012928112.1 | 5 - 6 | 1336 | 65 | |
XM_021259358.1 | XP_021115017.1 | 4 - 5 | 1293 | 65 | ||
XM_021259359.1 | XP_021115018.1 | 4 - 5 | 1217 | 65 | ||
XM_021259360.1 | XP_021115019.1 | 4 - 5 | 1294 | 65 | ||
XM_021259361.1 | XP_021115020.1 | 4 - 5 | 1293 | 65 | ||
XM_021259362.1 | XP_021115021.1 | 4 - 5 | 1293 | 65 | ||
XM_021259363.1 | XP_021115022.1 | 4 - 5 | 1293 | 65 | ||
XM_021259364.1 | XP_021115023.1 | 4 - 5 | 1294 | 65 | ||
XM_021259365.1 | XP_021115024.1 | 4 - 5 | 1294 | 65 | ||
XM_021259366.1 | XP_021115025.1 | 4 - 5 | 1294 | 65 | ||
XM_021259367.1 | XP_021115026.1 | 4 - 5 | 1294 | 65 | ||
XM_021259368.1 | XP_021115027.1 | 4 - 5 | 1294 | 65 | ||
XM_021259369.1 | XP_021115028.1 | 4 - 5 | 1293 | 65 | ||
XM_021259370.1 | XP_021115029.1 | 4 - 5 | 1294 | 65 | ||
XM_021259371.1 | XP_021115030.1 | 4 - 5 | 1294 | 65 | ||
XM_021259372.1 | XP_021115031.1 | 4 - 5 | 2231 | 65 | ||
XM_021259373.1 | XP_021115032.1 | 4 - 5 | 2229 | 65 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
