Assay Details
| Gene Symbol: Tmprss11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021262283.1 | XP_021117942.1 | 3 - 4 | 412 | 75 | |
| XM_021262284.1 | XP_021117943.1 | 3 - 4 | 413 | 75 | ||
| XM_021262286.1 | XP_021117945.1 | 3 - 4 | 409 | 75 | ||
More Information
Set Membership: |
|

Tmprss11a
Genomic Map