Assay Details
Gene Symbol: Ccdc191 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004835695.3 | XP_004835752.1 | 9 - 10 | 1553 | 60 | |
XM_021263437.1 | XP_021119096.1 | 9 - 10 | 1553 | 60 | ||
XM_021263441.1 | XP_021119100.1 | 9 - 10 | 1430 | 60 | ||
XM_021263442.1 | XP_021119101.1 | 6 - 7 | 985 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
