Assay Details
| Gene Symbol: LOC101717595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838454.2 | XP_004838511.2 | 12 - 13 | 1659 | 112 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101717595| Gene Symbol: LOC101717595 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004838454.2 | XP_004838511.2 | 12 - 13 | 1659 | 112 | |
Set Membership: |
|