Assay Details
Gene Symbol: LOC101717595 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838454.2 | XP_004838511.2 | 7 - 8 | 1167 | 79 | |
XM_021252763.1 | XP_021108422.1 | 7 - 8 | 1167 | 79 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
