Assay Details
Gene Symbol: Zdhhc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843036.3 | XP_004843093.1 | 10 - 11 | 1587 | 55 | |
XM_013071551.2 | XP_012927005.1 | 10 - 11 | 1569 | 55 | ||
XM_013071552.2 | XP_012927006.1 | 10 - 11 | 1563 | 55 | ||
XM_013071553.2 | XP_012927007.1 | 10 - 11 | 1527 | 55 | ||
XM_013071554.2 | XP_012927008.1 | 10 - 11 | 1518 | 55 | ||
XM_013071555.2 | XP_012927009.1 | 10 - 11 | 1587 | 55 | ||
XM_013071556.2 | XP_012927010.1 | 10 - 11 | 1432 | 55 | ||
XM_013071559.2 | XP_012927013.1 | 10 - 11 | 1433 | 55 | ||
XM_021256866.1 | XP_021112525.1 | 9 - 10 | 1540 | 55 | ||
XM_021256867.1 | XP_021112526.1 | 10 - 11 | 1492 | 55 | ||
XM_021256868.1 | XP_021112527.1 | 10 - 11 | 1457 | 55 | ||
XM_021256870.1 | XP_021112529.1 | 8 - 9 | 1193 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
