Assay Details
Gene Symbol: Pnpla1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846687.2 | XP_004846744.1 | 1 - 2 | 569 | 55 | |
XM_021259921.1 | XP_021115580.1 | 1 - 2 | 569 | 55 | ||
XM_021259922.1 | XP_021115581.1 | 1 - 2 | 569 | 55 | ||
XM_021259923.1 | XP_021115582.1 | 1 - 2 | 569 | 55 | ||
XM_021259924.1 | XP_021115583.1 | 1 - 2 | 569 | 55 | ||
XM_021259926.1 | XP_021115585.1 | 1 - 2 | 569 | 55 | ||
XM_021259927.1 | XP_021115586.1 | 1 - 2 | 569 | 55 | ||
XM_021259928.1 | XP_021115587.1 | 1 - 2 | 569 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
