Assay Details
| Gene Symbol: Cd101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854125.2 | XP_004854182.1 | 4 - 5 | 1409 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cd101| Gene Symbol: Cd101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854125.2 | XP_004854182.1 | 4 - 5 | 1409 | 64 | |
Set Membership: |
|