Assay Details
Gene Symbol: LOC101717715 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004859426.2 | XP_004859483.1 | 5 - 6 | 967 | 85 | |
XM_004859427.2 | XP_004859484.1 | 5 - 6 | 967 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() |
