Assay Details
| Gene Symbol: Tnfrsf11a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013077262.2 | XP_012932716.1 | 1 - 2 | 102 | 62 | |
| XM_021239256.1 | XP_021094915.1 | 1 - 2 | 79 | 62 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021239256.1 transcripts.
Set Membership: |
|

Tnfrsf11a
Genomic Map