Assay Details
Gene Symbol: Prr9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021249310.1 | XP_021104969.1 | 2 - 3 | 630 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021249310.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
