Assay Details
| Gene Symbol: Pwwp2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836457.3 | XP_004836514.1 | 1 - 2 | 678 | 120 | |
| XM_004836458.2 | XP_004836515.1 | 1 - 2 | 673 | 120 | ||
| XM_004836459.2 | XP_004836516.1 | 1 - 2 | 673 | 120 | ||
| XM_004836460.2 | XP_004836517.1 | 1 - 2 | 672 | 120 | ||
| XM_004836465.2 | XP_004836522.1 | 1 - 2 | 673 | 120 | ||
| XM_021240358.1 | XP_021096017.1 | 1 - 2 | 673 | 120 | ||
More Information
Set Membership: |
|

Pwwp2a
Genomic Map