Assay Details
| Gene Symbol: Fbxl17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004841762.3 | XP_004841819.1 | 5 - 6 | 1722 | 77 | |
| XM_021255850.1 | XP_021111509.1 | 6 - 7 | 1806 | 77 | ||
More Information
Set Membership: |
|

Fbxl17
Genomic Map