Assay Details
| Gene Symbol: LOC101725754 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843076.3 | XP_004843133.1 | 3 - 4 | 511 | 92 | |
| XM_004843077.3 | XP_004843134.1 | 3 - 4 | 512 | 92 | ||
| XM_013071580.2 | XP_012927034.1 | 3 - 4 | 513 | 92 | ||
More Information
Set Membership: |
|

LOC101725754
Genomic Map