Assay Details
| Gene Symbol: LOC101697345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846844.3 | XP_004846901.2 | 1 - 2 | 143 | 69 | |
| XM_021260035.1 | XP_021115694.1 | 1 - 2 | 143 | 69 | ||
| XM_021260036.1 | XP_021115695.1 | 1 - 2 | 143 | 69 | ||
More Information
Set Membership: |
|

LOC101697345
Genomic Map