Assay Details
Gene Symbol: Klf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848164.3 | XP_004848221.1 | 2 - 3 | 585 | 92 | |
XM_013073390.2 | XP_012928844.1 | 2 - 3 | 585 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
