Assay Details
Gene Symbol: Nat9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004860597.2 | XP_004860654.1 | 3 - 4 | 367 | 62 | |
XM_021241005.1 | XP_021096664.1 | 3 - 4 | 303 | 62 | ||
XM_021241006.1 | XP_021096665.1 | 3 - 4 | 303 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
