Assay Details
Gene Symbol: Lonrf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004872148.2 | XP_004872205.1 | 10 - 11 | 3034 | 59 | |
XM_004872149.2 | XP_004872206.1 | 9 - 10 | 2911 | 59 | ||
XM_013068277.2 | XP_012923731.1 | 11 - 12 | 3185 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
