Assay Details
Gene Symbol: LOC101725249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021245389.1 | XP_021101048.1 | 10 - 11 | 1394 | 100 | |
XM_021245391.1 | XP_021101050.1 | 10 - 11 | 1394 | 100 | ||
XM_021245392.1 | XP_021101051.1 | 10 - 11 | 1394 | 100 | ||
XM_021245393.1 | XP_021101052.1 | 9 - 10 | 1267 | 100 | ||
XM_021245394.1 | XP_021101053.1 | 8 - 9 | 1178 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
