Assay Details
Gene Symbol: LOC101725249 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021245389.1 | XP_021101048.1 | 5 - 6 | 733 | 83 | |
XM_021245391.1 | XP_021101050.1 | 5 - 6 | 733 | 83 | ||
XM_021245392.1 | XP_021101051.1 | 5 - 6 | 733 | 83 | ||
XM_021245393.1 | XP_021101052.1 | 4 - 5 | 606 | 83 | ||
XM_021245394.1 | XP_021101053.1 | 5 - 6 | 733 | 83 |
