Assay Details
Gene Symbol: LOC101726216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021254558.1 | XP_021110217.1 | 10 - 11 | 979 | 68 | |
XM_021254560.1 | XP_021110219.1 | 10 - 11 | 979 | 68 | ||
XM_021254561.1 | XP_021110220.1 | 10 - 11 | 979 | 68 | ||
XM_021254562.1 | XP_021110221.1 | 8 - 9 | 829 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
