Assay Details
Gene Symbol: LOC101726216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021254558.1 | XP_021110217.1 | 19 - 20 | 1798 | 103 | |
XM_021254560.1 | XP_021110219.1 | 18 - 19 | 1753 | 103 | ||
XM_021254561.1 | XP_021110220.1 | 16 - 17 | 1681 | 103 | ||
XM_021254562.1 | XP_021110221.1 | 17 - 18 | 1675 | 103 |

More Information
Set Membership: |
![]() ![]() ![]() |
