Assay Details
Gene Symbol: LOC101726216 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021254558.1 | XP_021110217.1 | 6 - 7 | 604 | 93 | |
XM_021254560.1 | XP_021110219.1 | 6 - 7 | 604 | 93 | ||
XM_021254561.1 | XP_021110220.1 | 6 - 7 | 604 | 93 | ||
XM_021254562.1 | XP_021110221.1 | 4 - 5 | 454 | 93 |
