Assay Details
| Gene Symbol: Spata21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263413.1 | XP_021119072.1 | 2 - 3 | 180 | 56 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Spata21| Gene Symbol: Spata21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021263413.1 | XP_021119072.1 | 2 - 3 | 180 | 56 | |
Set Membership: |
|