Assay Details
| Gene Symbol: Pcbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836219.2 | XP_004836276.2 | 2 - 3 | 212 | 65 | |
| XM_013077988.2 | XP_012933442.2 | 2 - 3 | 212 | 65 | ||
More Information
Set Membership: |
|

Pcbd2
Genomic Map