Assay Details
| Gene Symbol: Pcbd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004836219.2 | XP_004836276.2 | 3 - 4 | 344 | 100 | |
| XM_004836220.2 | XP_004836277.1 | 2 - 3 | 215 | 100 | ||
| XM_021238630.1 | XP_021094289.1 | 2 - 3 | 485 | 100 | ||
More Information
Set Membership: |
|

Pcbd2
Genomic Map