Assay Details
| Gene Symbol: Map3k7cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842252.3 | XP_004842309.1 | 4 - 5 | 521 | 99 | |
| XM_013071417.2 | XP_012926871.1 | 5 - 6 | 522 | 99 | ||
| XM_021256140.1 | XP_021111799.1 | 2 - 3 | 271 | 99 | ||
More Information
Set Membership: |
|

Map3k7cl
Genomic Map