Assay Details
| Gene Symbol: Cdh9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848670.2 | XP_004848727.1 | 11 - 12 | 2208 | 65 | |
| XM_021261533.1 | XP_021117192.1 | 5 - 6 | 1436 | 65 | ||
More Information
Set Membership: |
|

Cdh9
Genomic Map