Assay Details
Gene Symbol: Ccnyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851145.2 | XP_004851202.1 | 8 - 9 | 1083 | 74 | |
XM_004851146.2 | XP_004851203.1 | 8 - 9 | 991 | 74 | ||
XM_004851147.2 | XP_004851204.1 | 6 - 7 | 933 | 74 | ||
XM_004851148.2 | XP_004851205.1 | 9 - 10 | 831 | 74 | ||
XM_013074534.2 | XP_012929988.1 | 9 - 10 | 868 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
