Assay Details
| Gene Symbol: Acad8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862673.2 | XP_004862730.2 | 3 - 4 | 621 | 69 | |
| XM_021242780.1 | XP_021098439.1 | 3 - 4 | 621 | 69 | ||
| XM_021242781.1 | XP_021098440.1 | 3 - 4 | 621 | 69 | ||
More Information
Set Membership: |
|

Acad8
Genomic Map