Assay Details
Gene Symbol: Hrasls5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004874432.3 | XP_004874489.1 | 1 - 2 | 479 | 88 | |
XM_004874433.2 | XP_004874490.1 | 1 - 2 | 479 | 88 | ||
XM_021251859.1 | XP_021107518.1 | 2 - 3 | 536 | 88 |
