Assay Details
Gene Symbol: Erich6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021253123.1 | XP_021108782.1 | 11 - 12 | 1611 | 67 | |
XM_021253128.1 | XP_021108787.1 | 10 - 11 | 1487 | 67 | ||
XM_021253132.1 | XP_021108791.1 | 10 - 11 | 1146 | 67 | ||
XM_021253138.1 | XP_021108797.1 | 7 - 8 | 1320 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
