Assay Details
Gene Symbol: Rbbp8nl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021255214.1 | XP_021110873.1 | 2 - 3 | 264 | 63 | |
XM_021255215.1 | XP_021110874.1 | 2 - 3 | 264 | 63 | ||
XM_021255216.1 | XP_021110875.1 | 1 - 2 | 40 | 63 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021255216.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
