Assay Details
Gene Symbol: Hrasls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834766.2 | XP_004834823.1 | 1 - 2 | 180 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004834766.2 transcripts.
Set Membership: |
![]() ![]() |
