Assay Details
Gene Symbol: Hrasls | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004834766.2 | XP_004834823.1 | 3 - 4 | 591 | 76 | |
XM_004834767.3 | XP_004834824.1 | 3 - 4 | 510 | 76 | ||
XM_004834768.2 | XP_004834825.2 | 3 - 4 | 519 | 76 | ||
XM_021255579.1 | XP_021111238.1 | 3 - 4 | 594 | 76 | ||
XM_021255582.1 | XP_021111241.1 | 4 - 5 | 631 | 76 | ||
XM_021255586.1 | XP_021111245.1 | 3 - 4 | 627 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
