Assay Details
Gene Symbol: Bid | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837594.3 | XP_004837651.1 | 4 - 5 | 602 | 72 | |
XM_004837595.3 | XP_004837652.1 | 4 - 5 | 513 | 72 | ||
XM_004837596.3 | XP_004837653.1 | 4 - 5 | 872 | 72 | ||
XM_021249549.1 | XP_021105208.1 | 4 - 5 | 832 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
