Assay Details
| Gene Symbol: Pacsin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846740.3 | XP_004846797.1 | 2 - 3 | 876 | 54 | |
| XM_004846741.2 | XP_004846798.1 | 2 - 3 | 299 | 54 | ||
| XM_004846743.3 | XP_004846800.1 | 2 - 3 | 143 | 54 | ||
| XM_004846744.2 | XP_004846801.1 | 2 - 3 | 271 | 54 | ||
| XM_021259969.1 | XP_021115628.1 | 2 - 3 | 191 | 54 | ||
More Information
Set Membership: |
|

Pacsin1
Genomic Map