Assay Details
Gene Symbol: Mylk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847783.2 | XP_004847840.1 | 3 - 4 | 5383 | 84 | |
XM_021260752.1 | XP_021116411.1 | 3 - 4 | 5384 | 84 | ||
XM_021260753.1 | XP_021116412.1 | 4 - 5 | 715 | 84 | ||
XM_021260754.1 | XP_021116413.1 | 4 - 5 | 475 | 84 | ||
XM_021260755.1 | XP_021116414.1 | 3 - 4 | 648 | 84 | ||
XM_021260757.1 | XP_021116416.1 | 1 - 2 | 772 | 84 | ||
XM_021260758.1 | XP_021116417.1 | 2 - 3 | 5294 | 84 | ||
XM_021260759.1 | XP_021116418.1 | 2 - 3 | 2074 | 84 | ||
XM_021260760.1 | XP_021116419.1 | 3 - 4 | 5384 | 84 | ||
XM_021260761.1 | XP_021116420.1 | 3 - 4 | 5384 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004847783.2, XM_021260752.1, XM_021260753.1, XM_021260754.1, XM_021260755.1, XM_021260757.1, XM_021260758.1, XM_021260759.1, XM_021260760.1, XM_021260761.1 transcripts.
Set Membership: |
![]() ![]() |
Panther Classification:
