Assay Details
Gene Symbol: Mylk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847783.2 | XP_004847840.1 | 7 - 8 | 6478 | 84 | |
XM_021260752.1 | XP_021116411.1 | 7 - 8 | 6479 | 84 | ||
XM_021260753.1 | XP_021116412.1 | 8 - 9 | 1810 | 84 | ||
XM_021260754.1 | XP_021116413.1 | 8 - 9 | 1570 | 84 | ||
XM_021260755.1 | XP_021116414.1 | 7 - 8 | 1743 | 84 | ||
XM_021260757.1 | XP_021116416.1 | 5 - 6 | 1867 | 84 | ||
XM_021260758.1 | XP_021116417.1 | 6 - 7 | 6389 | 84 | ||
XM_021260759.1 | XP_021116418.1 | 6 - 7 | 3169 | 84 | ||
XM_021260760.1 | XP_021116419.1 | 7 - 8 | 6479 | 84 | ||
XM_021260762.1 | XP_021116421.1 | 5 - 6 | 1513 | 84 | ||
XM_021260763.1 | XP_021116422.1 | 5 - 6 | 1348 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
