Assay Details
Gene Symbol: Mylk4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847783.2 | XP_004847840.1 | 9 - 10 | 6674 | 73 | |
XM_021260752.1 | XP_021116411.1 | 9 - 10 | 6675 | 73 | ||
XM_021260753.1 | XP_021116412.1 | 10 - 11 | 2006 | 73 | ||
XM_021260754.1 | XP_021116413.1 | 10 - 11 | 1766 | 73 | ||
XM_021260755.1 | XP_021116414.1 | 9 - 10 | 1939 | 73 | ||
XM_021260757.1 | XP_021116416.1 | 7 - 8 | 2063 | 73 | ||
XM_021260758.1 | XP_021116417.1 | 8 - 9 | 6585 | 73 | ||
XM_021260759.1 | XP_021116418.1 | 8 - 9 | 3365 | 73 | ||
XM_021260760.1 | XP_021116419.1 | 9 - 10 | 6675 | 73 | ||
XM_021260761.1 | XP_021116420.1 | 9 - 10 | 6612 | 73 | ||
XM_021260762.1 | XP_021116421.1 | 7 - 8 | 1709 | 73 | ||
XM_021260763.1 | XP_021116422.1 | 7 - 8 | 1544 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
