Assay Details
Gene Symbol: Cep85l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004860164.3 | XP_004860221.2 | 5 - 6 | 1223 | 59 | |
XM_013077928.2 | XP_012933382.1 | 5 - 6 | 1223 | 59 | ||
XM_021240679.1 | XP_021096338.1 | 5 - 6 | 1223 | 59 | ||
XM_021240680.1 | XP_021096339.1 | 5 - 6 | 1223 | 59 | ||
XM_021240681.1 | XP_021096340.1 | 4 - 5 | 1508 | 59 | ||
XM_021240683.1 | XP_021096342.1 | 5 - 6 | 1223 | 59 | ||
XM_021240684.1 | XP_021096343.1 | 5 - 6 | 1223 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() |
