Assay Details
| Gene Symbol: Prss22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864660.3 | XP_004864717.1 | 3 - 4 | 421 | 103 | |
| XM_013065537.2 | XP_012920991.1 | 3 - 4 | 419 | 103 | ||
More Information
Set Membership: |
|

Prss22
Genomic Map