Assay Details
Gene Symbol: LOC101705383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004868063.3 | XP_004868120.1 | 14 - 15 | 2234 | 83 | |
XM_013066756.2 | XP_012922210.1 | 14 - 15 | 2170 | 83 | ||
XM_021247070.1 | XP_021102729.1 | 14 - 15 | 2655 | 83 | ||
XM_021247071.1 | XP_021102730.1 | 15 - 16 | 2293 | 83 | ||
XM_021247072.1 | XP_021102731.1 | 14 - 15 | 2513 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
