Assay Details
Gene Symbol: LOC101705383 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004868063.3 | XP_004868120.1 | 8 - 9 | 1754 | 70 | |
XM_013066756.2 | XP_012922210.1 | 8 - 9 | 1690 | 70 | ||
XM_021247070.1 | XP_021102729.1 | 8 - 9 | 2175 | 70 | ||
XM_021247071.1 | XP_021102730.1 | 9 - 10 | 1813 | 70 | ||
XM_021247072.1 | XP_021102731.1 | 8 - 9 | 2033 | 70 | ||
XM_021247073.1 | XP_021102732.1 | 8 - 9 | 1755 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
