Assay Details
| Gene Symbol: LOC101699122 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021250483.1 | XP_021106142.1 | 1 - 2 | 207 | 68 | |
| XM_021250484.1 | XP_021106143.1 | 1 - 2 | 208 | 68 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021250483.1, XM_021250484.1 transcripts.
Set Membership: |
|

LOC101699122
Genomic Map