Assay Details
| Gene Symbol: Gpatch11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004839296.2 | XP_004839353.1 | 6 - 7 | 673 | 60 | |
| XM_021253725.1 | XP_021109384.1 | 6 - 7 | 746 | 60 | ||
More Information
Set Membership: |
|

Gpatch11
Genomic Map