Assay Details
Gene Symbol: LOC101711512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845532.3 | XP_004845589.1 | 10 - 11 | 2544 | 63 | |
XM_004845533.3 | XP_004845590.1 | 9 - 10 | 2406 | 63 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
