Assay Details
| Gene Symbol: LOC101711512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845532.3 | XP_004845589.1 | 12 - 13 | 2731 | 93 | |
| XM_004845533.3 | XP_004845590.1 | 11 - 12 | 2593 | 93 | ||
More Information
Set Membership: |
|

LOC101711512
Genomic Map