Assay Details
| Gene Symbol: LOC101711512 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004845532.3 | XP_004845589.1 | 28 - 29 | 5100 | 87 | |
| XM_004845533.3 | XP_004845590.1 | 27 - 28 | 4962 | 87 | ||
More Information
Set Membership: |
|

LOC101711512
Genomic Map