Assay Details
Gene Symbol: LOC101711512 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845532.3 | XP_004845589.1 | 2 - 3 | 1218 | 93 | |
XM_004845533.3 | XP_004845590.1 | 2 - 3 | 1218 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
